The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168620.1 | NP_001162091.1 | 3 - 4 | 1092 | 115 |
NM_032003.2 | NP_114392.1 | 3 - 4 | 1178 | 115 | |
GenBank mRNA | AF233377.1 | - | 3 - 4 | 1155 | 115 |
AK032341.1 | - | 3 - 4 | 1081 | 115 | |
AK220510.1 | - | 3 - 4 | 1121 | 115 | |
BC138977.1 | - | 3 - 4 | 1093 | 115 | |
BC138978.1 | - | 3 - 4 | 1093 | 115 |