Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pde6c
Entrez Gene ID
110855
Gene Name
phosphodiesterase 6C, cGMP specific, cone, alpha prime
Gene Aliases
cpfl1
Chromosome Location
Chr.19: 38132781 - 38183955 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170959.1 NP_001164430.1 15 - 16 2095 75
NM_033614.2 NP_291092.1 15 - 16 2095 75
GenBank mRNA AF411063.1 - 15 - 16 1930 75
BC027050.1 - 15 - 16 2095 75

View Full Product Details