Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim7
Entrez Gene ID
94089
Gene Name
tripartite motif-containing 7
Gene Aliases
AI790312
Chromosome Location
Chr.11: 48826138 - 48850644 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF220033.1 - 1 - 2 586 52
BC138188.1 - 1 - 2 562 52

View Full Product Details