Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acss1
Entrez Gene ID
68738
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
1110032O15Rik, AI788978, Acas2, Acas2l, AceCS2
Chromosome Location
Chr.2: 150618111 - 150668240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080575.2 NP_542142.1 4 - 5 832 106
GenBank mRNA AB046742.1 - 4 - 5 821 106
AK088244.1 - 4 - 5 811 106
AK146568.1 - 4 - 5 833 106
AK158119.1 - 4 - 5 827 106
AK172412.1 - 4 - 5 818 106
AK172470.1 - 4 - 5 807 106
AK173275.1 - 4 - 5 793 106
BC092278.1 - 4 - 5 861 106

View Full Product Details