Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plin2
Entrez Gene ID
11520
Gene Name
perilipin 2
Gene Aliases
AA407157, ADPH, Adfp, Adrp
Chromosome Location
Chr.4: 86626910 - 86670059 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007408.3 NP_031434.3 6 - 7 916 62
GenBank mRNA AK011014.1 - 6 - 7 905 62
AK078967.1 - 5 - 6 892 62
AK090277.1 - 6 - 7 848 62
AK150177.1 - 6 - 7 853 62
AK152081.1 - 6 - 7 852 62
AK152263.1 - 6 - 7 844 62
AK152877.1 - 6 - 7 852 62
AK152896.1 - 6 - 7 844 62
AK152982.1 - 6 - 7 876 62
AK153222.1 - 6 - 7 874 62
AK154582.1 - 6 - 7 853 62
AK167140.1 - 6 - 7 854 62
AY035850.1 - 5 - 6 775 62
AY035851.1 - 5 - 6 775 62
BC054766.1 - 6 - 7 829 62
M93275.1 - 6 - 7 853 62

View Full Product Details