Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Npdc1
Entrez Gene ID
18146
Gene Name
neural proliferation, differentiation and control 1
Gene Aliases
AI314472, NPDC-1
Chromosome Location
Chr.2: 25403050 - 25409494 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008721.4 NP_032747.2 5 - 6 846 97
GenBank mRNA AK146466.1 - 5 - 6 735 97
BC003320.1 - 5 - 6 813 97
BC023887.1 - 4 - 5 880 97
L03814.1 - 5 - 6 650 97
X67209.1 - 5 - 6 650 97

View Full Product Details