The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008916.2 | NP_032942.1 | 8 - 9 | 1101 | 63 |
GenBank mRNA | AF483522.1 | - | 8 - 9 | 1021 | 63 |
AF483523.1 | - | 8 - 9 | 1021 | 63 | |
AK052817.1 | - | 7 - 8 | 1845 | 63 | |
AK054436.1 | - | 8 - 9 | 1120 | 63 | |
AK078104.1 | - | 8 - 9 | 1112 | 63 | |
AK164290.1 | - | 8 - 9 | 1090 | 63 | |
BC014725.1 | - | 8 - 9 | 1031 | 63 | |
BC066112.1 | - | 8 - 9 | 1101 | 63 | |
U96724.1 | - | 8 - 9 | 1132 | 63 |