The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164677.1 | NP_001158149.1 | 8 - 9 | 1220 | 66 |
NM_001164678.1 | NP_001158150.1 | 8 - 9 | 1205 | 66 | |
NM_011052.2 | NP_035182.2 | 8 - 9 | 1205 | 66 | |
GenBank mRNA | AF119955.1 | - | 8 - 9 | 1166 | 66 |
AJ005073.1 | - | 8 - 9 | 1053 | 66 | |
AK029827.1 | - | 8 - 9 | 1172 | 66 | |
AK031256.1 | - | 8 - 9 | 1206 | 66 | |
AK033943.1 | - | 8 - 9 | 1194 | 66 | |
AK129340.1 | - | 8 - 9 | 1175 | 66 | |
AK145893.1 | - | 8 - 9 | 1193 | 66 | |
AK147736.1 | - | 8 - 9 | 1178 | 66 | |
AK167574.1 | - | 8 - 9 | 1163 | 66 | |
AK169704.1 | - | 8 - 9 | 1206 | 66 | |
BC026823.1 | - | 8 - 9 | 1147 | 66 | |
BC051123.1 | - | 8 - 9 | 1056 | 66 |