Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pld3
Entrez Gene ID
18807
Gene Name
phospholipase D family, member 3
Gene Aliases
Sam-9
Chromosome Location
Chr.7: 27531645 - 27553213 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011116.1 NP_035246.1 4 - 5 440 64
GenBank mRNA AF026124.1 - 4 - 5 440 64
BC068170.1 - 4 - 5 437 64
BC076586.1 - 4 - 5 504 64

View Full Product Details