Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdcd7
Entrez Gene ID
50996
Gene Name
programmed cell death 7
Gene Aliases
C80112, ES18
Chromosome Location
Chr.9: 65346068 - 65359643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016688.2 NP_057897.2 3 - 4 1315 67
GenBank mRNA AF083929.1 - 3 - 4 1309 67
AK004096.1 - 1 - 2 108 67
AK136141.1 - 1 - 2 94 67
AK155050.1 - 3 - 4 1280 67
BC022772.1 - 2 - 3 352 67
DV656988.1 - 2 - 3 328 67

View Full Product Details