Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Barhl1
Entrez Gene ID
54422
Gene Name
BarH-like 1 (Drosophila)
Gene Aliases
Dres115, MBH2
Chromosome Location
Chr.2: 28907680 - 28918415 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164186.1 NP_001157658.1 3 - 4 1152 67
NM_019446.4 NP_062319.1 2 - 3 1435 67
GenBank mRNA AB043980.1 - 2 - 3 1450 67
AF264026.1 - 4 - 5 1210 67
AJ237590.1 - 2 - 3 710 67
AK036312.1 - 2 - 3 887 67
AK048063.1 - 2 - 3 288 67
BC055731.1 - 3 - 4 1153 67

View Full Product Details