Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arpc3
Entrez Gene ID
56378
Gene Name
actin related protein 2/3 complex, subunit 3
Gene Aliases
1110006A04Rik, 21kDa, AA408672, AI788639, p21-Ar, p21Arc
Chromosome Location
Chr.5: 122391928 - 122406178 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB038243.1 - 4 - 5 379 77
AK003485.1 - 5 - 6 474 77
AK005437.1 - 5 - 6 462 77
BC054440.1 - 5 - 6 410 77
BC055036.1 - 5 - 6 410 77

View Full Product Details