Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klk11
Entrez Gene ID
56538
Gene Name
kallikrein related-peptidase 11
Gene Aliases
2310015I08Rik, Prss20, TLSP
Chromosome Location
Chr.7: 43774617 - 43779263 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177373.1 NP_001170844.1 2 - 3 264 65
NM_019974.2 NP_064358.1 1 - 2 230 65
GenBank mRNA AB016226.1 - 2 - 3 114 65
AB016227.1 - 2 - 3 224 65
AK009360.1 - 2 - 3 265 65
AK009659.1 - 2 - 3 240 65
AK009720.1 - 2 - 3 265 65
BC115518.2 - 2 - 3 96 65

View Full Product Details