The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271476.1 | NP_001258405.1 | 5 - 6 | 1804 | 58 |
NM_001271477.1 | NP_001258406.1 | 4 - 5 | 1667 | 58 | |
NM_001271479.1 | NP_001258408.1 | 3 - 4 | 1358 | 58 | |
NM_021319.5 | NP_067294.2 | 4 - 5 | 1696 | 58 | |
NR_073184.1 | 4 - 5 | 1569 | 58 | ||
GenBank mRNA | AB084140.1 | - | 5 - 6 | 1801 | 58 |
AB084141.1 | - | 5 - 6 | 1714 | 58 | |
AF149837.1 | - | 4 - 5 | 1693 | 58 | |
AF149838.1 | - | 4 - 5 | 1606 | 58 | |
AF315578.1 | - | 3 - 4 | 1355 | 58 | |
AF315584.1 | - | 4 - 5 | 1566 | 58 | |
AF317548.1 | - | 5 - 6 | 1562 | 58 | |
AK050019.1 | - | 4 - 5 | 1668 | 58 | |
AY282722.1 | - | 4 - 5 | 1634 | 58 | |
BC019396.1 | - | 4 - 5 | 1669 | 58 |