Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekho1
Entrez Gene ID
67220
Gene Name
pleckstrin homology domain containing, family O member 1
Gene Aliases
2810052M02Rik, CKIP-1, Ckip1, JZA-20, Jza2
Chromosome Location
Chr.3: 95988809 - 95995863 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023320.2 NP_075809.1 5 - 6 563 71
GenBank mRNA AF168675.1 - 5 - 6 563 71
AK012945.1 - 5 - 6 72
AK014374.1 - 5 - 6 583 71
AK170025.1 - 5 - 6 678 71
AK170922.1 - 5 - 6 671 71
BC117003.1 - 5 - 6 570 71
BC125441.1 - 5 - 6 570 71

View Full Product Details