The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164290.1 | NP_001157762.1 | 9 - 10 | 920 | 62 |
NM_001164291.1 | NP_001157763.1 | 8 - 9 | 812 | 62 | |
NM_001164292.1 | NP_001157764.1 | 8 - 9 | 809 | 62 | |
NM_001164293.1 | NP_001157765.1 | 8 - 9 | 779 | 62 | |
NM_001164294.1 | NP_001157766.1 | 8 - 9 | 758 | 62 | |
NM_001164295.1 | NP_001157767.1 | 8 - 9 | 755 | 62 | |
NM_024192.2 | NP_077154.2 | 9 - 10 | 911 | 62 | |
GenBank mRNA | AB232359.1 | - | 7 - 8 | 710 | 62 |
AK003457.1 | - | 8 - 9 | 714 | 62 | |
AK013884.1 | - | 8 - 9 | 770 | 62 | |
BC003442.1 | - | 9 - 10 | 875 | 62 |