The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025617.2 | NP_079893.1 | 3 - 4 | 409 | 59 |
GenBank mRNA | AK008715.1 | - | 3 - 4 | 364 | 59 |
AK011991.1 | - | 3 - 4 | 350 | 59 | |
BC028321.1 | - | 3 - 4 | 347 | 59 |