The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025806.2 | NP_080082.1 | 4 - 5 | 775 | 74 |
GenBank mRNA | AK003169.1 | - | 4 - 5 | 697 | 74 |
AK170196.1 | - | 4 - 5 | 760 | 74 | |
AK172021.1 | - | 4 - 5 | 675 | 74 | |
BC019793.1 | - | 4 - 5 | 727 | 74 |