The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025862.2 | NP_080138.2 | 6 - 7 | 756 | 82 |
GenBank mRNA | AK009379.1 | - | 6 - 7 | 735 | 82 |
AK019502.1 | - | 6 - 7 | 753 | 82 | |
AK029897.1 | - | 6 - 7 | 720 | 82 | |
AK049908.1 | - | 2 - 3 | 1076 | 82 | |
AK051091.1 | - | 6 - 7 | 711 | 82 | |
AK077335.1 | - | 6 - 7 | 736 | 82 | |
AK161255.1 | - | 6 - 7 | 710 | 82 | |
AK178437.1 | - | 3 - 4 | 292 | 82 | |
AK207169.1 | - | 3 - 4 | 244 | 82 | |
BC026535.1 | - | 6 - 7 | 730 | 82 | |
BC037644.1 | - | 6 - 7 | 732 | 82 |