Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Qpctl
Entrez Gene ID
67369
Gene Name
glutaminyl-peptide cyclotransferase-like
Gene Aliases
1810019P04Rik, BB101812, gQC
Chromosome Location
Chr.7: 19140217 - 19149196 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026111.3 NP_080387.2 4 - 5 799 114
GenBank mRNA AK007555.1 - 4 - 5 844 114
AK007701.1 - 4 - 5 827 114
AK035587.1 - 4 - 5 799 114
AK045687.1 - 4 - 5 803 114
BC058181.1 - 4 - 5 822 114

View Full Product Details