Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fer
Entrez Gene ID
14158
Gene Name
fer (fms/fps related) protein kinase
Gene Aliases
AV082135, C330004K01Rik, Fert, Fert2
Chromosome Location
Chr.17: 63829950 - 64146519 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037997.3 NP_001033086.2 18 - 19 2467 66
NM_001286415.1 NP_001273344.1 17 - 18 2295 66
NM_008000.2 NP_032026.2 13 - 14 1749 66
GenBank mRNA AK082799.1 - 15 - 16 1960 66
BC051249.1 - 10 - 11 1211 66
BC058100.1 - 17 - 18 2295 66
M32054.1 - 13 - 14 1749 66
U76762.1 - 17 - 18 2400 66

View Full Product Details