The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008564.2 | NP_032590.2 | 1 - 2 | 138 | 63 |
GenBank mRNA | AK037751.1 | - | 1 - 2 | 62 | 63 |
AK077801.1 | - | 1 - 2 | 39 | 63 | |
AK088156.1 | - | 1 - 2 | 51 | 63 | |
AK088378.1 | - | 1 - 2 | 38 | 63 | |
AK146187.1 | - | 1 - 2 | 53 | 63 | |
AK146375.1 | - | 1 - 2 | 62 | 63 | |
BC055318.1 | - | 1 - 2 | 47 | 63 | |
BY324067.1 | - | 1 - 2 | 138 | 63 | |
D86725.1 | - | 1 - 2 | 58 | 63 |