The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008571.1 | NP_032597.1 | 4 - 5 | 626 | 75 |
GenBank mRNA | AK133521.1 | - | 4 - 5 | 627 | 75 |
AY007570.1 | - | 4 - 5 | 626 | 75 | |
BC119248.1 | - | 4 - 5 | 619 | 75 | |
BC119250.1 | - | 4 - 5 | 619 | 75 |