Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map2
Entrez Gene ID
17756
Gene Name
microtubule-associated protein 2
Gene Aliases
G1-397-34, MAP-2, Mtap-2, Mtap2, repro4
Chromosome Location
Chr.1: 66175246 - 66442583 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA M21041.1 - 8 - 9 4616 79

View Full Product Details