Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fos
Entrez Gene ID
14281
Gene Name
FBJ osteosarcoma oncogene
Gene Aliases
D12Rfj1, c-fos, cFos
Chromosome Location
Chr.12: 85473901 - 85477269 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010234.2 NP_034364.1 2 - 3 527 67
GenBank mRNA AK154418.1 - 2 - 3 539 67
AY423854.1 - 1 - 2 199 67
BC029814.1 - 2 - 3 527 67

View Full Product Details