Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mad1l1
Entrez Gene ID
17120
Gene Name
MAD1 mitotic arrest deficient 1-like 1
Gene Aliases
AI173502, AW550425, Mad1
Chromosome Location
Chr.5: 140008689 - 140321591 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010752.3 NP_034882.2 8 - 9 981 78
GenBank mRNA AF083812.1 - 8 - 9 988 78
AF083813.1 - 8 - 9 988 78
AK136981.1 - 8 - 9 984 78
BC010702.1 - 8 - 9 979 78

View Full Product Details