The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035228.2 | NP_001030305.1 | 6 - 7 | 1098 | 76 |
NM_011375.3 | NP_035505.2 | 6 - 7 | 1310 | 76 | |
GenBank mRNA | AB013302.1 | - | 5 - 6 | 849 | 76 |
AB018048.1 | - | 6 - 7 | 1038 | 76 | |
AF119416.1 | - | 6 - 7 | 1088 | 76 | |
AK012961.1 | - | 6 - 7 | 1079 | 76 | |
AK158594.1 | - | 6 - 7 | 1131 | 76 | |
AK159000.1 | - | 6 - 7 | 1087 | 76 | |
AK165726.1 | - | 6 - 7 | 1100 | 76 | |
BC138557.1 | - | 6 - 7 | 1015 | 76 | |
BC138559.1 | - | 6 - 7 | 1013 | 76 | |
Y15003.1 | - | 6 - 7 | 1071 | 76 |