Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sox5
Entrez Gene ID
20678
Gene Name
SRY (sex determining region Y)-box 5
Gene Aliases
A730017D01Rik, AI528773
Chromosome Location
Chr.6: 143828425 - 144781989 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113559.2 NP_001107031.2 11 - 12 1656 125
NM_001243163.1 NP_001230092.1 11 - 12 1548 125
NM_011444.3 NP_035574.2 12 - 13 1800 125
GenBank mRNA AB006330.1 - 12 - 13 1712 125
AJ010604.1 - 11 - 12 1538 125
AK015212.1 - 5 - 6 609 125
AK160154.1 - 5 - 6 614 125
BC110478.2 - 11 - 12 1540 125
KF793916.1 - 12 - 13 1501 125
X65657.1 - 5 - 6 585 125

View Full Product Details