The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113559.2 | NP_001107031.2 | 12 - 13 | 1826 | 70 |
NM_001243163.1 | NP_001230092.1 | 12 - 13 | 1718 | 70 | |
NM_011444.3 | NP_035574.2 | 13 - 14 | 1970 | 70 | |
GenBank mRNA | AB006330.1 | - | 13 - 14 | 1882 | 70 |
AJ010604.1 | - | 12 - 13 | 1708 | 70 | |
AK160154.1 | - | 6 - 7 | 784 | 70 | |
BC110478.2 | - | 12 - 13 | 1710 | 70 | |
KF793916.1 | - | 13 - 14 | 1671 | 70 | |
X65657.1 | - | 6 - 7 | 755 | 70 | |
X65658.1 | - | 1 - 2 | 72 | 70 |