The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011521.2 | NP_035651.1 | 4 - 5 | 469 | 55 |
GenBank mRNA | AK134326.1 | - | 4 - 5 | 467 | 55 |
AK153445.1 | - | 4 - 5 | 469 | 55 | |
AK159215.1 | - | 4 - 5 | 468 | 55 | |
AK164952.1 | - | 4 - 5 | 468 | 55 | |
BC005679.1 | - | 4 - 5 | 454 | 55 | |
D89571.1 | - | 4 - 5 | 469 | 55 |