The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011936.2 | NP_036066.2 | 5 - 6 | 1017 | 81 |
GenBank mRNA | AJ237917.1 | - | 5 - 6 | 1003 | 81 |
AK040866.1 | - | 5 - 6 | 995 | 81 | |
AK045465.1 | - | 5 - 6 | 994 | 81 | |
AK046935.1 | - | 5 - 6 | 976 | 81 | |
AK049502.1 | - | 5 - 6 | 1010 | 81 | |
AK088881.1 | - | 5 - 6 | 1003 | 81 | |
AK129437.1 | - | 5 - 6 | 984 | 81 | |
AK161060.1 | - | 5 - 6 | 976 | 81 | |
BC022222.1 | - | 5 - 6 | 968 | 81 | |
BC057008.1 | - | 5 - 6 | 983 | 81 |