The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163704.1 | NP_001157176.1 | 1 - 2 | 311 | 73 |
GenBank mRNA | AF176526.1 | - | 1 - 2 | 210 | 73 |
AK144715.1 | - | 1 - 2 | 248 | 73 | |
AK150612.1 | - | 1 - 2 | 279 | 73 | |
AK152888.1 | - | 1 - 2 | 161 | 73 | |
BC017512.1 | - | 1 - 2 | 107 | 73 |