The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016926.1 | NP_058622.1 | 17 - 18 | 2533 | 71 |
GenBank mRNA | AF172722.1 | - | 17 - 18 | 2533 | 71 |
AK086398.1 | - | 17 - 18 | 2534 | 71 | |
AK129067.1 | - | 16 - 17 | 2908 | 71 | |
AK216500.1 | - | 1 - 2 | 98 | 71 | |
BC057156.1 | - | 17 - 18 | 2524 | 71 |