The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019519.2 | NP_062392.1 | 6 - 7 | 1044 | 71 |
GenBank mRNA | AF127656.1 | - | 7 - 8 | 892 | 71 |
AF127657.1 | - | 6 - 7 | 835 | 71 | |
AF127658.1 | - | 6 - 7 | 1044 | 71 | |
AF127659.1 | - | 6 - 7 | 1044 | 71 | |
AF127660.1 | - | 7 - 8 | 892 | 71 | |
AF127661.1 | - | 6 - 7 | 835 | 71 | |
AF127662.1 | - | 7 - 8 | 963 | 71 | |
AK002625.1 | - | 7 - 8 | 826 | 71 | |
AK146917.1 | - | 7 - 8 | 915 | 71 | |
BC012214.1 | - | 7 - 8 | 897 | 71 |