The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019791.2 | NP_062765.1 | 3 - 4 | 953 | 94 |
GenBank mRNA | AB029448.1 | - | 3 - 4 | 953 | 94 |
AF319975.1 | - | 2 - 3 | 741 | 94 | |
AK017275.1 | - | 3 - 4 | 871 | 94 | |
AK049660.1 | - | 3 - 4 | 904 | 94 | |
AK087937.1 | - | 3 - 4 | 904 | 94 | |
AK132248.1 | - | 3 - 4 | 892 | 94 | |
AK135452.1 | - | 3 - 4 | 903 | 94 | |
AK145872.1 | - | 3 - 4 | 873 | 94 | |
AK146867.1 | - | 3 - 4 | 903 | 94 | |
AK146908.1 | - | 3 - 4 | 903 | 94 | |
AK220150.1 | - | 4 - 5 | 810 | 94 | |
BC016438.1 | - | 3 - 4 | 869 | 94 |