Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Srd5a3
- Entrez Gene ID
-
57357
- Gene Name
- steroid 5 alpha-reductase 3
- Gene Aliases
-
1110025P14Rik, A430076C09, AV364670, AW987574, D730040M03Rik, H5ar, S5AR 3, Srd5a2l
- Chromosome Location
-
Chr.5: 76140273 - 76155503 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC145647.1 |
- |
4 - 5 |
650 |
64 |
View Full Product Details