The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022322.2 | NP_071717.1 | 2 - 3 | 384 | 65 |
GenBank mRNA | AB055422.1 | - | 2 - 3 | 191 | 65 |
AF191768.1 | - | 2 - 3 | 330 | 65 | |
AF219993.1 | - | 2 - 3 | 257 | 65 | |
AF291655.1 | - | 2 - 3 | 409 | 65 | |
AK003748.1 | - | 2 - 3 | 270 | 65 | |
BC006919.1 | - | 2 - 3 | 193 | 65 | |
BC049944.1 | - | 2 - 3 | 384 | 65 |