Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrps22
Entrez Gene ID
64655
Gene Name
mitochondrial ribosomal protein S22
Gene Aliases
3100002P07Rik, Rpms22
Chromosome Location
Chr.9: 98588730 - 98601679 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025485.3 NP_079761.1 3 - 4 539 113
GenBank mRNA AK003836.1 - 2 - 3 307 113
AK013925.1 - 3 - 4 520 113
AK154600.1 - 3 - 4 520 113
BC051198.1 - 3 - 4 519 113

View Full Product Details