Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crisp2
Entrez Gene ID
22024
Gene Name
cysteine-rich secretory protein 2
Gene Aliases
CRISP-2, GAPDL5, Tpx-1, Tpx1
Chromosome Location
Chr.17: 40764734 - 40794146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009420.2 NP_033446.1 2 - 3 202 98
GenBank mRNA AK018856.1 - 2 - 3 133 98
BC049615.1 - 2 - 3 154 98
M25533.1 - 2 - 3 154 98

View Full Product Details