Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsx
Entrez Gene ID
22127
Gene Name
testis specific X-linked gene
Gene Aliases
AV207184
Chromosome Location
Chr.X: 103409101 - 103424588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009440.2 NP_033466.1 4 - 5 334 121
GenBank mRNA BC048493.1 - 4 - 5 334 121
X99796.1 - 4 - 5 319 121

View Full Product Details