The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009513.2 | NP_033539.1 | 3 - 4 | 421 | 96 |
GenBank mRNA | AK138191.1 | - | 3 - 4 | 352 | 96 |
AK153741.1 | - | 2 - 3 | 329 | 96 | |
AK160941.1 | - | 3 - 4 | 407 | 96 | |
BC052019.1 | - | 2 - 3 | 244 | 96 | |
D83206.1 | - | 3 - 4 | 374 | 96 |