Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfx
Entrez Gene ID
22764
Gene Name
zinc finger protein X-linked
Gene Aliases
Zfx5, Zfx5,6, Zfx6
Chromosome Location
Chr.X: 94074631 - 94150643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044386.1 NP_001037851.1 9 - 9 2054 75
NM_011768.2 NP_035898.2 8 - 8 1986 75
GenBank mRNA AK141652.1 - 1 - 1 316 75
BC137859.1 - 7 - 7 1654 75
BC144979.1 - 6 - 6 1504 75
M32308.1 - 9 - 9 1762 75
M32309.1 - 8 - 8 1935 75

Gene Symbol
Zfa-ps
Entrez Gene ID
22639
Gene Name
zinc finger protein, autosomal, pseudogene
Gene Aliases
Zfa, Zfx
Chromosome Location
Chr.10: 52542320 - 52545739 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_037920.1 1 - 1 1951 75
GenBank mRNA AK029509.1 - 1 - 1 1951 75

View Full Product Details