The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009577.3 | NP_033603.2 | 3 - 4 | 405 | 80 |
GenBank mRNA | AK146692.1 | - | 3 - 4 | 399 | 80 |
AK159077.1 | - | 3 - 4 | 391 | 80 | |
BC050942.1 | - | 3 - 4 | 363 | 80 | |
BC067196.1 | - | 3 - 4 | 363 | 80 | |
U69133.1 | - | 3 - 4 | 403 | 80 |