The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010072.3 | NP_034202.1 | 2 - 3 | 283 | 78 |
GenBank mRNA | AB004789.1 | - | 2 - 3 | 257 | 78 |
AI099209.1 | - | 2 - 3 | 283 | 78 | |
AK004834.1 | - | 2 - 3 | 276 | 78 | |
AK005268.1 | - | 2 - 3 | 269 | 78 | |
BC061151.1 | - | 2 - 3 | 263 | 78 |