The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010267.3 | NP_034397.1 | 5 - 6 | 791 | 80 |
GenBank mRNA | AK045460.1 | - | 5 - 6 | 791 | 80 |
AK083814.1 | - | 5 - 6 | 792 | 80 | |
AK139057.1 | - | 5 - 6 | 634 | 80 | |
AK149281.1 | - | 5 - 6 | 802 | 80 | |
BC048177.1 | - | 5 - 6 | 767 | 80 | |
BC051135.1 | - | 5 - 6 | 756 | 80 | |
BC060212.1 | - | 5 - 6 | 764 | 80 | |
Y17850.1 | - | 5 - 6 | 779 | 80 |