The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010321.1 | NP_034451.1 | 5 - 6 | 725 | 99 |
GenBank mRNA | AK007398.1 | - | 5 - 6 | 551 | 99 |
BC014283.1 | - | 5 - 6 | 724 | 99 | |
D89664.1 | - | 5 - 6 | 725 | 99 |