The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252266.1 | NP_001239195.1 | 11 - 12 | 1331 | 69 |
NM_010492.3 | NP_034622.3 | 13 - 14 | 1807 | 69 | |
GenBank mRNA | AB303567.1 | - | 12 - 13 | 1318 | 69 |
BC002030.1 | - | 13 - 14 | 1441 | 69 | |
CD775505.1 | - | 1 - 2 | 69 | ||
CO044997.1 | - | 1 - 2 | 69 | ||
U26459.1 | - | 13 - 14 | 1430 | 69 | |
U26460.1 | - | 12 - 13 | 1336 | 69 | |
U26461.1 | - | 11 - 12 | 1272 | 69 | |
U34595.1 | - | 11 - 12 | 1303 | 69 |