The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011678.2 | NP_035808.2 | 19 - 20 | 2623 | 74 |
GenBank mRNA | AK143582.1 | - | 19 - 20 | 2558 | 74 |
AK149964.1 | - | 19 - 20 | 2581 | 74 | |
AK163201.1 | - | 2 - 3 | 269 | 74 | |
AK164245.1 | - | 10 - 11 | 1398 | 74 | |
AK169372.1 | - | 20 - 21 | 2602 | 74 | |
AK169933.1 | - | 19 - 20 | 2593 | 74 | |
AK171271.1 | - | 19 - 20 | 2593 | 74 | |
AK171633.1 | - | 19 - 20 | 2576 | 74 | |
BC011341.1 | - | 18 - 19 | 2400 | 74 | |
L00681.1 | - | 19 - 20 | 2586 | 74 |