Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbl2
Entrez Gene ID
27368
Gene Name
transducin (beta)-like 2
Gene Aliases
C76179, WS-bTRP
Chromosome Location
Chr.5: 135149426 - 135163876 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013763.2 NP_038791.2 5 - 6 738 85
GenBank mRNA AF097523.1 - 5 - 6 725 85
AK010899.1 - 5 - 6 758 85
AK088198.1 - 4 - 5 612 85
AK132366.1 - 5 - 6 764 85
AK165686.1 - 5 - 6 758 85
BC039273.1 - 5 - 6 771 85
BC048185.1 - 5 - 6 738 85

View Full Product Details