Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il27ra
Entrez Gene ID
50931
Gene Name
interleukin 27 receptor, alpha
Gene Aliases
CRL1, IL-27R, Tccr, WSX-1, Wsx1, zcytor1
Chromosome Location
Chr.8: 84030286 - 84042629 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016671.3 NP_057880.2 1 - 2 129 84
GenBank mRNA AF053005.1 - 1 - 2 84
AF265243.1 - 1 - 2 84
AK133753.1 - 1 - 2 144 84
BC032878.1 - 1 - 2 84

View Full Product Details